Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010578.2 | NP_034708.1 | 6 | 1028 | ||
GenBank mRNA | AK076526.1 | - | 2 | 325 | ||
AK088016.1 | - | 6 | 929 | |||
AK088729.1 | - | 6 | 775 | |||
AK135521.1 | - | 5 | 704 | |||
AK159689.1 | - | 6 | 870 | |||
AK167682.1 | - | 6 | 926 | |||
BC050906.1 | - | 6 | 901 | 5721348 | ||
X15202.1 | - | 5 | 721 | |||
Y00769.1 | - | 6 | 816 |