Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oc90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010953.2 | NP_035083.1 | 13 | 1097 | ||
GenBank mRNA | AF091846.1 | - | 10 | 906 | ||
AF093591.1 | - | 13 | 1207 | |||
AK012981.1 | - | 13 | 1099 | |||
BC048716.1 | - | 11 | 968 | 6334417 |