Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011516.2 | NP_035646.2 | 2 | 225 | ||
GenBank mRNA | AK015000.1 | - | 2 | 226 | ||
AK133229.1 | - | 2 | 226 | |||
BC137967.1 | - | 2 | 187 | 8860986 | ||
L41069.1 | - | 2 | 206 | |||
Z38118.1 | - | 2 | 216 |
Gene Symbol: Sycp1-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015899.1 | - | 1 | 97 |