Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019419.2 | NP_062292.1 | 6 | 1028 | ||
GenBank mRNA | AF172088.1 | - | 6 | 1014 | ||
AF223953.1 | - | 6 | 965 | |||
AK090001.1 | - | 6 | 980 | |||
AK150042.1 | - | 6 | 981 | |||
AK150507.1 | - | 6 | 979 | |||
AK151046.1 | - | 4 | 746 | |||
AK151272.1 | - | 6 | 978 | |||
AK151507.1 | - | 6 | 978 | |||
AK152565.1 | - | 6 | 978 | |||
AK152885.1 | - | 6 | 975 | |||
AK154644.1 | - | 6 | 981 | |||
AK167872.1 | - | 6 | 983 | |||
AK167888.1 | - | 6 | 1031 | |||
AK167912.1 | - | 6 | 978 | |||
AK168172.1 | - | 6 | 981 | |||
AK168245.1 | - | 6 | 980 | |||
AK169191.1 | - | 6 | 978 | |||
BC010196.1 | - | 6 | 968 | 3157834 | ||
BC046792.1 | - | 6 | 951 | 5717527 |