Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019428.3 | NP_062301.3 | 5 | 316 | ||
GenBank mRNA | AK004137.1 | - | 5 | 314 | ||
AK160536.1 | - | 5 | 243 | |||
AK169253.1 | - | 5 | 243 | |||
BC062146.1 | - | 5 | 301 | 6518236 | ||
CN722047.1 | - | 5 | 363 | |||
U95123.1 | - | 5 | 295 |