Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019501.3 | NP_062374.2 | 4 | 511 | ||
GenBank mRNA | AB210841.1 | - | 4 | 431 | ||
AF118855.1 | - | 2 | 206 | |||
AK012307.1 | - | 4 | 439 | |||
BC026820.1 | - | 4 | 413 | 4039464 | ||
BC107273.1 | - | 4 | 326 | |||
BC107274.1 | - | 4 | 330 |