Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003719.2 | NP_001003719.1 | 7 | 1321 | ||
NM_001112714.2 | NP_001106185.1 | 7 | 1321 | |||
NM_001286263.1 | NP_001273192.1 | 7 | 1321 | |||
NM_019994.5 | NP_064378.4 | 7 | 1321 | |||
GenBank mRNA | AK042116.1 | - | 7 | 887 | ||
AK046680.1 | - | 7 | 973 | |||
AK129236.1 | - | 7 | 775 | |||
AK153664.1 | - | 7 | 884 | |||
AY596972.1 | - | 7 | 972 | |||
AY596973.1 | - | 7 | 972 | |||
AY596974.1 | - | 7 | 972 | |||
BC145119.1 | - | 7 | 902 | 9053667 | ||
BC150707.1 | - | 7 | 902 | 9087618 |