Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020514.2 | NP_065260.2 | 1 | 509 | ||
GenBank mRNA | BC119195.1 | - | 1 | 509 | 8733944 | |
BC119199.1 | - | 1 | 509 | 8733948 |