Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021607.3 | NP_067620.3 | 4 | 453 | ||
GenBank mRNA | AF240469.1 | - | 4 | 403 | ||
AK033314.1 | - | 4 | 395 | |||
AK129102.1 | - | 4 | 453 | |||
AK170173.1 | - | 4 | 399 | |||
AK170244.1 | - | 4 | 418 | |||
AK170369.1 | - | 4 | 442 | |||
AK172579.1 | - | 4 | 398 | |||
BC019998.1 | - | 4 | 398 | 4458780 | ||
CD549803.1 | - | 4 | 453 |