Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021607.3 | NP_067620.3 | 7 | 845 | ||
GenBank mRNA | AF240469.1 | - | 7 | 795 | ||
AK033314.1 | - | 7 | 788 | |||
AK129102.1 | - | 7 | 845 | |||
AK170173.1 | - | 7 | 791 | |||
AK170244.1 | - | 7 | 810 | |||
AK170369.1 | - | 7 | 834 | |||
AK172579.1 | - | 7 | 790 | |||
BC019998.1 | - | 7 | 790 | 4458780 |