Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023230.2 | NP_075719.1 | 4 | 489 | ||
GenBank mRNA | AK002551.1 | - | 4 | 451 | ||
AK012021.1 | - | 4 | 468 | |||
AK158230.1 | - | 4 | 489 | |||
AK170413.1 | - | 4 | 460 | |||
BC003449.1 | - | 4 | 476 | 2654197 | ||
BC019372.1 | - | 3 | 330 | 2646198 | ||
BC092253.1 | - | 4 | 455 | 5055396 |