Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024433.2 | NP_077753.1 | 8 | 1997 | ||
GenBank mRNA | AK005064.1 | - | 8 | 1944 | ||
AK159515.1 | - | 1 | 690 | |||
AK167319.1 | - | 8 | 1998 | |||
BC003858.1 | - | 8 | 1941 | 3485422 |