Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025414.3 | NP_079690.2 | 21 | 2546 | ||
GenBank mRNA | AK018609.1 | - | 6 | 576 | ||
AK080117.1 | - | 19 | 2265 | |||
BC007156.1 | - | 14 | 1462 | 3486558 | ||
BC046294.1 | - | 20 | 2274 | |||
BC060115.1 | - | 18 | 3991 |