Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025414.3 | NP_079690.2 | 25 | 3588 | ||
GenBank mRNA | AK018609.1 | - | 10 | 1618 | ||
AK080117.1 | - | 23 | 3307 | |||
BC007156.1 | - | 18 | 2504 | 3486558 | ||
BC046294.1 | - | 24 | 3311 | |||
BC060115.1 | - | 22 | 5033 |