Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285435.1 | NP_001272364.1 | 8 | 823 | ||
GenBank mRNA | AK008355.1 | - | 8 | 788 | ||
AK008364.1 | - | 8 | 788 | |||
AK008440.1 | - | 8 | 787 | |||
AK028819.1 | - | 8 | 1295 | |||
AK029081.1 | - | 8 | 764 | |||
AK154947.1 | - | 5 | 413 | |||
BC034843.1 | - | 6 | 676 | |||
BC058577.1 | - | 4 | 1399 | |||
CK624091.1 | - | 3 | 328 |