Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291860.1 | NP_001278789.1 | 24 | 3191 | ||
NM_005529.6 | NP_005520.4 | 24 | 3188 | |||
XM_011541318.2 | XP_011539620.1 | 25 | 3177 | |||
XM_017001120.1 | XP_016856609.1 | 25 | 3318 | |||
XM_017001121.1 | XP_016856610.1 | 24 | 3267 | |||
XM_017001122.1 | XP_016856611.1 | 24 | 3264 | |||
GenBank mRNA | M64283.1 | - | 1 | 57 | ||
M85289.1 | - | 24 | 3188 | |||
S76436.1 | - | 3 | 441 | |||
X62515.1 | - | 24 | 3154 |