Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026316.2 | NP_080592.2 | 10 | 1710 | ||
GenBank mRNA | AF362571.1 | - | 9 | 1632 | ||
AK004004.1 | - | 3 | 729 | |||
AK005615.1 | - | 4 | 840 | |||
AK154500.1 | - | 10 | 1756 | |||
AK159447.1 | - | 10 | 1724 | |||
AK160106.1 | - | 10 | 1725 | |||
BC023305.1 | - | 10 | 1732 | |||
BC046597.1 | - | 10 | 1710 | 6312211 | ||
BC082792.1 | - | 3 | 748 |