Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027268.2 | NP_081544.1 | 2, 3 | 301 | ||
GenBank mRNA | AK012765.1 | - | 2, 3 | 329 | ||
AK032573.1 | - | 2, 3 | 328 | |||
AK050481.1 | - | 2, 3 | 319 | |||
AK082953.1 | - | 2, 3 | 301 | |||
AK089968.1 | - | 2, 3 | 291 | |||
AK129084.1 | - | 2, 3 | 288 | |||
AK145904.1 | - | 2, 3 | 303 | |||
BC023889.1 | - | 2, 3 | 288 | 5252046 |