Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206567.1 | NP_001193496.1 | 4 | 1027 | ||
NM_005531.2 | NP_005522.2 | 5 | 1195 | |||
XM_005245127.3 | XP_005245184.1 | 6 | 1512 | |||
XM_006711290.1 | XP_006711353.1 | 5 | 1195 | |||
XM_017001149.1 | XP_016856638.1 | 6 | 1512 | |||
XM_017001150.1 | XP_016856639.1 | 5 | 1195 | |||
GenBank mRNA | AB208989.1 | - | 5 | 1165 | ||
AF208043.1 | - | 5 | 1169 | |||
AK296228.1 | - | 4 | 910 | |||
AK307957.1 | - | 5 | 976 | |||
AY138863.1 | - | 4 | 905 | |||
BC017059.1 | - | 5 | 1162 | 3914632 | ||
M63838.1 | - | 5 | 1169 |