Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027268.2 | NP_081544.1 | 6 | 910 | ||
GenBank mRNA | AK012765.1 | - | 6 | 938 | ||
AK032573.1 | - | 6 | 937 | |||
AK050481.1 | - | 6 | 928 | |||
AK082953.1 | - | 6 | 910 | |||
AK089968.1 | - | 6 | 900 | |||
AK129084.1 | - | 6 | 897 | |||
AK145904.1 | - | 6 | 912 | |||
BC023889.1 | - | 6 | 897 | 5252046 |