Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027554.2 | NP_081830.2 | 3 | 1330 | ||
GenBank mRNA | AK028904.1 | - | 3 | 1309 | ||
AK049287.1 | - | 3 | 1298 | |||
AK052219.1 | - | 3 | 1353 | |||
AK054116.1 | - | 3 | 1332 | |||
AK129464.1 | - | 3 | 1286 | |||
AK139639.1 | - | 3 | 1343 | |||
AK142200.1 | - | 3 | 1325 | |||
BC054404.1 | - | 3 | 1330 | 6494448 | ||
BC057122.1 | - | 3 | 1320 | 6834543 | ||
BC058784.1 | - | 3 | 1258 | 6310341 | ||
BC066786.1 | - | 3 | 1274 |