Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290546.1 | NP_001277475.1 | 5 | 617 | ||
NM_027642.2 | NP_081918.1 | 7 | 903 | |||
GenBank mRNA | AK016480.1 | - | 7 | 904 | ||
AK028498.1 | - | 5 | 539 | |||
AK038320.1 | - | 7 | 823 | |||
AK122560.1 | - | 7 | 833 | |||
AK141804.1 | - | 7 | 846 | |||
AK163345.1 | - | 7 | 857 | |||
BC043127.1 | - | 7 | 851 | 6410361 | ||
BC055330.1 | - | 7 | 846 | 6406483 | ||
BC057374.1 | - | 7 | 809 | 6826091 |