Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027998.4 | NP_082274.1 | 1 | 1322 | ||
GenBank mRNA | AK009330.1 | - | 1 | 1243 | ||
AK037108.1 | - | 1 | 1277 | |||
AK145246.1 | - | 1 | 1326 | |||
BC019534.1 | - | 1 | 1307 | 4207924 | ||
BC085262.1 | - | 1 | 1307 | 6396088 |