Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029457.3 | NP_083733.1 | 6 | 638 | ||
GenBank mRNA | AF368904.1 | - | 6 | 684 | ||
AK015987.1 | - | 6 | 617 | |||
AK031030.1 | - | 6 | 623 | |||
AY188288.1 | - | 5 | 410 | |||
BC031652.1 | - | 6 | 613 | 3483046 |