Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029457.3 | NP_083733.1 | 12 | 1374 | ||
GenBank mRNA | AF368904.1 | - | 12 | 1420 | ||
AK015987.1 | - | 12 | 1353 | |||
AK031030.1 | - | 12 | 1359 | |||
AY188288.1 | - | 11 | 1146 | |||
BC031652.1 | - | 12 | 1349 | 3483046 | ||
BC040805.1 | - | 4 | 1318 | |||
BC042514.1 | - | 4 | 1318 |