Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029619.2 | NP_083895.1 | 4 | 428 | ||
GenBank mRNA | AK009912.1 | - | 4 | 419 | ||
BC021619.1 | - | 4 | 428 | 5150285 |