Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030697.2 | NP_109622.1 | 4, 5 | 1457 | ||
GenBank mRNA | AK002623.1 | - | 4, 5 | 1458 | ||
AK132011.1 | - | 4, 5 | 1479 |