Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290699.1 | NP_001277628.1 | 3 | 410 | ||
NM_001290700.1 | NP_001277629.1 | 3 | 410 | |||
NM_133817.3 | NP_598578.1 | 3 | 410 | |||
GenBank mRNA | AK015797.1 | - | 3 | 385 | ||
AK030088.1 | - | 3 | 243 | |||
AK122541.1 | - | 3 | 235 | |||
AK210231.1 | - | 2 | 157 | |||
BC019546.1 | - | 3 | 246 | 4038791 | ||
BC062154.1 | - | 3 | 200 | 6331597 |