Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144874.4 | NP_659123.2 | 2 | 353 | ||
GenBank mRNA | AK048099.1 | - | 2 | 335 | ||
AK051792.1 | - | 2 | 251 | |||
AK141631.1 | - | 2 | 285 | |||
BC011509.1 | - | 2 | 313 | 3981950 | ||
BC021498.1 | - | 2 | 273 |