Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005542.4 | NP_005533.2 | 6 | 1215 | ||
NM_198336.2 | NP_938150.2 | 6 | 830 | |||
NM_198337.2 | NP_938151.1 | 4 | 923 | |||
XM_005249542.1 | XP_005249599.1 | 7 | 1286 | |||
XM_005249543.1 | XP_005249600.1 | 7 | 1301 | |||
XM_017012177.1 | XP_016867666.1 | 6 | 1230 | |||
GenBank mRNA | AK095977.1 | - | 4 | 493 | ||
AK291675.1 | - | 6 | 1087 | |||
BC001880.1 | - | 6 | 1153 | 3546928 | ||
BQ059075.1 | - | 6 | 820 |