Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145491.2 | NP_663466.2 | 5 | 2774 | ||
GenBank mRNA | AB060650.1 | - | 5 | 2592 | ||
BC048813.1 | - | 5 | 2584 | 2582017 | ||
BC056363.1 | - | 5 | 2774 | 6821501 | ||
BC057451.1 | - | 5 | 2307 |