Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005547.2 | NP_005538.2 | 2 | 342 | ||
GenBank mRNA | AK300492.1 | - | 358 | |||
AK301649.1 | - | 224 | ||||
BC046391.2 | - | 1 | 315 | 4749952 | ||
BG679040.1 | - | 2 | 342 | |||
BM811601.1 | - | 62 |