Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146367.2 | NP_666479.1 | 1 | 429 | ||
GenBank mRNA | AK135940.1 | - | 2 | 526 | ||
BC119350.1 | - | 1 | 429 | 8734099 | ||
BC120621.1 | - | 1 | 429 | 40129544 |