Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005555.3 | NP_005546.2 | 9 | 2132 | ||
GenBank mRNA | BC034535.1 | - | 9 | 2131 | 4754558 |