Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146727.2 | NP_666938.2 | 1 | 774 |
Gene Symbol: Olfr612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200027.1 | NP_001186956.1 | 1 | 774 |