Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146795.1 | NP_667006.1 | 1 | 519 | ||
GenBank mRNA | BC132194.1 | - | 1 | 541 | 40130471 | |
BC132196.1 | - | 1 | 541 | 40130473 |