Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185056.1 | NP_001171985.1 | 3 | 429 | ||
NM_005602.5 | NP_005593.2 | 3 | 759 | |||
GenBank mRNA | AJ245901.1 | - | 3 | 557 | ||
AK294087.1 | - | 3 | 400 | |||
AK312923.1 | - | 3 | 756 | |||
BC013577.1 | - | 3 | 747 | 3895040 | ||
BT019402.1 | - | 3 | 557 | |||
BT019403.1 | - | 3 | 557 | |||
DA258090.1 | - | 3 | 429 |