Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147100.3 | NP_667311.3 | 1 | 1822 | ||
GenBank mRNA | AK156691.1 | - | 1 | 1821 | ||
BC145889.1 | - | 1 | 434 | 40131192 |