Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005618.3 | NP_005609.3 | 2 | 568 | ||
XM_005266934.3 | XP_005266991.1 | 2 | 1034 | |||
GenBank mRNA | AF196571.1 | - | 2 | 416 | ||
AK314234.1 | - | 2 | 440 | |||
AY358892.1 | - | 2 | 135 | |||
BC028096.1 | - | 2 | 652 | |||
DR000029.1 | - | 2 | 187 | |||
EU927387.1 | - | 2 | 259 |