Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF536756.1 | - | 4 | 805 | ||
AK122497.2 | - | 4 | 701 | |||
BC138257.1 | - | 4 | 803 | 8861276 | ||
BC145214.1 | - | 4 | 803 |