Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163470.1 | NP_001156942.1 | 12 | 2352 | ||
NM_172275.2 | NP_758479.2 | 12 | 2340 | |||
GenBank mRNA | AA168469.1 | - | 1 | 34 | ||
AK003586.1 | - | 3 | 805 | |||
AK042299.1 | - | 4 | 998 | |||
AK150038.1 | - | 12 | 2321 | |||
BC021421.1 | - | 2 | 1098 | |||
BC023072.1 | - | 6 | 2756 | |||
BC038396.1 | - | 12 | 2296 | 3987227 |