Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130184.1 | NP_001123656.1 | 6 | 651 | ||
NM_001130185.1 | NP_001123657.1 | 6 | 731 | |||
NM_001130186.1 | NP_001123658.1 | 6 | 689 | |||
NM_172303.4 | NP_758507.3 | 6 | 749 | |||
GenBank mRNA | AK049332.1 | - | 6 | 654 | ||
AK129445.1 | - | 6 | 644 | |||
AK147466.1 | - | 6 | 611 | |||
BC020316.2 | - | 6 | 671 | 5250231 |