Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161823.1 | NP_001155295.1 | 3 | 383 | ||
NM_172382.2 | NP_759014.2 | 3 | 383 | |||
GenBank mRNA | AK054095.1 | - | 3 | 347 | ||
AK129187.1 | - | 3 | 238 | |||
AK145066.1 | - | 3 | 369 | |||
AK145947.1 | - | 3 | 305 | |||
BC028866.1 | - | 3 | 286 | 4207552 | ||
BY308642.1 | - | 3 | 384 |