Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161823.1 | NP_001155295.1 | 17 | 2620 | ||
NM_172382.2 | NP_759014.2 | 17 | 2620 | |||
GenBank mRNA | AK054095.1 | - | 17 | 2584 | ||
AK129187.1 | - | 17 | 2475 | |||
AK136085.1 | - | 8 | 1277 | |||
AK145066.1 | - | 17 | 2606 | |||
AK145947.1 | - | 17 | 2542 | |||
AK217187.1 | - | 4 | 368 | |||
BC028866.1 | - | 17 | 2523 | 4207552 |