Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172402.3 | NP_765990.2 | 7 | 1616 | ||
GenBank mRNA | AK011759.1 | - | 1 | 533 | ||
AK031187.1 | - | 7 | 1421 | |||
AK145178.1 | - | 7 | 1576 | |||
AK167501.1 | - | 7 | 1426 | |||
AK170643.1 | - | 7 | 1585 | |||
AK172029.1 | - | 7 | 1583 | |||
BC031874.1 | - | 7 | 1550 | 5149984 |