Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172410.2 | NP_765998.1 | 3 | 196 | ||
GenBank mRNA | AJ505051.1 | - | 2 | 132 | ||
AK010450.1 | - | 2 | 148 | |||
AK028864.1 | - | 3 | 197 | |||
AK129055.1 | - | 2 | 146 | |||
AK167105.1 | - | 2 | 147 | |||
AK169693.1 | - | 2 | 166 | |||
BC023140.1 | - | 2 | 142 | 3991335 |