Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172410.2 | NP_765998.1 | 6, 7 | 594 | ||
GenBank mRNA | AK010450.1 | - | 5, 6 | 545 | ||
AK028864.1 | - | 6, 7 | 595 | |||
AK045380.1 | - | 453 | ||||
AK129055.1 | - | 5, 6 | 544 | |||
AK167105.1 | - | 5, 6 | 545 | |||
AK169194.1 | - | 3, 4 | 315 | |||
AK169693.1 | - | 5, 6 | 564 | |||
BC023140.1 | - | 5, 6 | 540 | 3991335 |