Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159527.1 | NP_001152999.1 | 5 | 541 | ||
NM_172470.3 | NP_766058.3 | 5 | 541 | |||
GenBank mRNA | AK015490.1 | - | 5 | 506 | ||
AK029896.1 | - | 5 | 496 | |||
AK031061.1 | - | 5 | 532 | |||
AK053724.1 | - | 5 | 496 | |||
AK083971.1 | - | 5 | 486 | |||
AK173151.1 | - | 5 | 483 | |||
BC056925.1 | - | 5 | 650 | 5704780 |