Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201359.1 | NP_001188288.1 | 2, 3 | 192 | ||
NM_005624.3 | NP_005615.2 | 2, 3 | 192 | |||
XM_011528177.2 | XP_011526479.1 | 3, 4 | 393 | |||
XM_017027119.1 | XP_016882608.1 | 3, 4 | 359 | |||
XM_017027120.1 | XP_016882609.1 | 3, 4 | 288 | |||
XM_017027121.1 | XP_016882610.1 | 3, 4 | 234 | |||
XM_017027122.1 | XP_016882611.1 | 3, 4 | 371 | |||
GenBank mRNA | AB046579.1 | - | 1, 2 | 81 | ||
AK292662.1 | - | 2, 3 | 192 | |||
BC125136.1 | - | 1, 2 | 85 | 40119547 | ||
BC130561.1 | - | 1, 2 | 127 | 40146592 | ||
BC144463.1 | - | 1, 2 | 127 | 9052989 | ||
U86358.1 | - | 1, 2 | 81 |