Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159527.1 | NP_001152999.1 | 25 | 3214 | ||
NM_172470.3 | NP_766058.3 | 26 | 3247 | |||
GenBank mRNA | AK029896.1 | - | 25 | 3169 | ||
AK083971.1 | - | 25 | 3159 | |||
BC056925.1 | - | 26 | 3356 | 5704780 |